Would you like to log in again?
VWR Collection disposable garments: High quality performance products that offer superb value for money
Detailed technical data sheets available!
VWR Collection disposable garments: High quality performance products that offer superb value for money
Quality and savings go hand in hand!
  • Bouffant caps latex-free!
  • Hygiene masks: different colours, with ties or earloops, all latex-free
  • FFP2 masks with or without valve: State-of-the-art regarding comfort
  • Disposable overalls, frocks and laboratory coats, individually packed
Whether you are looking for disposable bouffant caps, hygiene masks, shoe covers, disposable laboratory coats or frocks, the VWR Collection is perfect to fit your demand and budget. For your everyday needs in your laboratory or production facility (pharmaceutical, chemical, semi-conductor, food and beverage, ...).
Description Packed Pk Cat. No. Price Quantity
Bouffant caps
Non woven, PP, white, 530 mm Bag 100 113-8241E
Non woven, PP, white, 530 mm Box 100 113-8244E
Non woven, PP, blue, 530 mm Box 100 113-8245E
Non woven, PP, green, 530 mm Box 100 113-8246E
Non woven, PP, white, 610 mm Bag 100 113-8243E
Non woven, PP, blue, 610 mm Bag 100 113-8288E
Non woven, PP, green, 610 mm Bag 100 113-8289E
Clip, white, 500 mm Bag 100 113-8250E
Clip, white, 500 mm Box 250 113-8247E
Clip, blue, 500 mm Box 250 113-8248E
Clip, green, 500 mm Box 250 113-8249E
Clip, red, 500 mm Box 100 113-8290E
Clip, yellow, 500 mm Box 100 113-8291E
Visor, PP, standard, blue, 510 mm Bag 100 113-8252E
Visor, PP, standard, white 510 mm Bag 100 113-8251E
Visor, PP, long hair, white, 510 mm Bag 100 113-8253E
Beard covers
PP, white Bag 100 113-8254E
Sleeve protectors
PP/PE, white, 200×400 mm(Ø×L) Bag 100 113-8255E
LDPE/LLDPE, white, 200×400 mm(Ø×L) Bag 100 113-8256E
LDPE/LLDPE, blue, 200×400 mm(Ø×L) Bag 100 113-8257E
Overalls
PP, white, tapered collar, zip, size L Bag 25 113-8258E
PP, white, tapered collar, zip, size XL Bag 25 113-8259E
PP, white, tapered collar, zip, size XXL Bag 25 113-8260E
PP, white, tapered collar, zip, size XXXL Bag 25 113-8292E
PP, white, hood, zip, size M Bag 25 113-8261E
PP, white, hood, zip, size L Bag 25 113-8262E
PP, white, hood, zip, size XL Bag 25 113-8263E
PP, white, hood, zip, size XXL Bag 25 113-8264E
PP, white, hood, zip, size XXXL Bag 25 113-8293E
SPP 50 g, white, hood, zip, ESD, size M Bag 50 113-8305E
SPP 50 g, white, hood, zip, ESD, size L Bag 50 113-8306E
SPP 50 g, white, hood, zip, ESD, size XL Bag 50 113-8307E
SPP 50 g, white, hood, zip, ESD, size XXL Bag 50 113-8308E
SPP 50 g, white, hood, zip, ESD, size XXXL Bag 50 113-8309E
SPP 50 g, white, hood, zip, ESD, size XXXXL Bag 50 113-8310E
SPP 40 g, white, collar, dual zipper, size M Bag 25 113-8311E
SPP 40 g, white, collar, dual zipper, size L Bag 25 113-8312E
SPP 40 g, white, collar, dual zipper, size XL Bag 25 113-8313E
SPP 40 g, white, collar, dual zipper, size XXL Bag 25 113-8314E
Smocks
Visitors, LDPE/LLDPE, white, hood, press studs, one size Bag 100 113-8265E
Labcoats
PP/PE, white, waterproof, press studs, size L Bag 30 113-8266E
PP/PE, white, waterproof, press studs, size XL Bag 30 113-8267E
PP/PE, white, waterproof, press studs, size XXL Bag 30 113-8268E
PP, white, elasticated wrists, press studs, size L - 30 113-8272E
PP, white, elasticated wrists, press studs, size XL - 30 113-8273E
PP, white, elasticated wrists, press studs, size XXL - 30 113-8274E
PP, white, elasticated wrists, zip, size XL (one size) - 30 113-8271E
PP, white, straight wrists, press studs, size L - 100 113-8275E
PP, white, straight wrists, press studs, size XL - 100 113-8276E
PP, white, straight wrists, press studs, size XXL - 100 113-8277E
SPP, 45 g, white, tailored collar, press studs, size M Bag 50 113-8297E
SPP, 45 g, white, tailored collar, press studs, size L Bag 50 113-8298E
SPP, 45 g, white, tailored collar, press studs, size XL Bag 50 113-8299E
SPP, 45 g, white, tailored collar, press studs, size XXL Bag 50 113-8302E
SPP, 45 g, white, tailored collar, press studs, size XXXL Bag 50 113-8303E
SPP, 45 g, white, tailored collar, press studs, size XXXXL Bag 50 113-8304E
Frocks
PP, white, no pockets, elasticated wrists, press studs, size XL Bag 30 113-8269E
PP, blue, no pockets, elasticated wrists, press studs, size L Bag 30 113-8295E
PP, blue, no pockets, elasticated wrists, press studs, size XL Bag 30 113-8296E
PP, white, no pockets, elasticated wrists, zip, size XL Bag 30 113-8270E
PP, white, no pockets, elasticated wrists, zip, size XL Bag 30 113-8294E
Aprons
LDPE/LLDPE, white, 700x1200 mm - 100 113-8278E
LDPE/LLDPE, white, 780x1400 mm - 100 113-8279E
Shoe covers
Non woven, white, 400x180 mm(LxH) - 200 113-8280E
CPE, blue, 360x150 mm(LxH) - 100 113-8281E
CPE, white, 360x150 mm(LxH) - 100 113-8284E
Non woven, CPE sole, white, 410x160 mm(LxH) - 200 113-8282E
Non woven, CPE sole, blue, 410x16 0 mm(LxH) - 200 113-8283E
Non woven, PP, laminated, blue, 430x170 mm(LxH) - 300 113-8285E
Non woven PP with latex pattern, 460x170 mm - 100 113-8286E
Non woven PP/PE, 400x170 mm - 300 113-8287E
Isolation gowns
SMS, 45 g, long sleeves, blue, 115x136 cm Bag 50 113-8316E
SMS, 45 g, long sleeves, green, 115x136 cm Bag 50 113-8317E
SMS, 45 g, long sleeves, pink, 115x136 cm Bag 50 113-8319E
SMS, 45 g, long sleeves, red, 115x136 cm Bag 50 113-8318E
SMS, 45 g, long sleeves, white, 115x136 cm Bag 50 113-8315E
SMS, 45 g, long sleeves, yellow, 115x136 cm Bag 50 113-8320E
SMS, 60 g, long sleeves, blue, 115x136 cm Bag 50 113-8322E
SMS, 60 g, long sleeves, green, 115x136 cm Bag 50 113-8323E
SMS, 60 g, long sleeves, pink, 115x136 cm Bag 50 113-8325E
SMS, 60 g, long sleeves, red, 115x136 cm Bag 50 113-8324E
SMS, 60 g, long sleeves, white, 115x136 cm Bag 50 113-8321E
SMS, 60 g, long sleeves, yellow, 115x136 cm Bag 50 113-8326E