Would you like to log in again?
VWR® cooling packs
VWR® cooling packs
VALID UNTIL: 31/10/2017
Designed to safeguard temperature-sensitive products during storage and transport.
Effective when used with insulated shipping containers.
  • Can be chilled in a freezer, refrigerator or low temperature incubator
  • Keeps their shape during all stages of use: Phase changes at 0 °C and returns to gel state after thawing
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals and biologicals
WxDxH Weight Pk Cat. No. Promo price Quantity
89x165x19 mm 227 g 72 216-0192E
140x152x25 mm 341 g 48 216-0193E
140x165x25 mm 454 g 36 216-0194E
203x140x32 mm 682 g 24 216-0195E
267x140x32 mm 909 g 18 216-0196E