Would you like to log in again?
VWR Collection cleanroom nitrile gloves
NEW!
VWR Collection cleanroom nitrile gloves
Switch cleanroom gloves from latex to nitrile with the brand new gloves from VWR Collection.
  • High tensile strength and strong permeation resistance against a wide variety of solvents, acids and bases
  • Double bagged in sealed polyethylene cleanroom bags (100 pieces/liner; 10 liners in a bag/carton). Sterile gloves individually packed (50 pairs in a liner, 4 liners in a bag/carton)
  • Extractables measured according to IEST-RP-CC005.3
  • Sterile version: Endotoxin level <20 EU/glove
  • Compliant to the PPE Directive (89/686/EEC) and EN 374
  • Contain no natural rubber latex
Sterile (ISO 5) and non sterile (ISO 4 and ISO 5) available. A 'clean' glove, quality tested according to ASTM, FTMS and IEST standards. Provide comfort, protection and tactile sensitivity with exceptional low particle counts.
Size Pk Cat. No. Price Quantity
Class 100 cleanroom nitrile gloves
X-Small 1000 112-4537E
Small 1000 112-4538E
Medium 1000 112-4539E
Large 1000 112-4540E
X-Large 1000 112-4541E
XX-Large 1000 112-4542E
Class 10 cleanroom nitrile gloves
X-Small 1000 112-4543E
Small 1000 112-4544E
Medium 1000 112-4545E
Large 1000 112-4546E
X-Large 1000 112-4547E
XX-Large 1000 112-4548E
Class 100 cleanroom nitrile gloves, sterile
6 200 112-4549E
6,5 200 112-4550E
7 200 112-4551E
7,5 200 112-4552E
8 200 112-4553E
8,5 200 112-4554E
9 200 112-4555E
10 200 112-4556E