Would you like to log in again?
VWR® cleanroom face masks
VWR® cleanroom face masks
VALID UNTIL: 31/10/2017
New 'duckbill' style
The VWR Collection range of  cleanroom face masks has recently been extended with the introduction of our new 'duckbill' style masks.
  • Superior particle and bacterial filtration efficiency whilst being extremely cost effective
  • Available as sterile or non sterile (sterile are individually bagged)
  • Hood loops, ear loops or ties with a malleable nose band
  • Natural rubber latex-free, glass fibre and silicon-free
  • Low particle shedding and moisture resistant
Packed 10 masks per bag, 20 bags per case.
Available to pre-order for delivery in October!
Duckbill face mask Pk Cat. No. Promo price Quantity
Sterile (individually packed)
With hood loop 200 113-8681E
With ear loop 200 113-8682E
With ties 200 113-8683E
Non sterile
With hood loop 200 113-8684E
With ear loop 200 113-8685E
With ties 200 113-8686E