Would you like to log in again?
Aqueous solutions for biopharmaceutical production
Order your aqueous solutions ready to use!
Aqueous solutions for biopharmaceutical production
These ready to use aqueous solutions for biopharmaceutical production are mixed in a cleanroom with pharmaceutical grade raw materials (Ph. Eur.) and purified water (Ph. Eur., USP).
  • Solutions prepared in a controlled environment
  • Comprehensive Certificate of Analysis
  • Typical specifications for products in biopharmaceutical applications
  • Low bioburden
VWR is a global chemical solution provider offering reliable and transparent supply chain solutions, while managing the ever increasing complexity that your business faces, by using our global strength.
Description Pack size Cat. No. Promo Price Quantity
Sodium hydroxide 0,1 mol*l 2.5l VWRC85519.320E
Sodium hydroxide 0,1 mol*l 25l VWRC85519.460E
Sodium hydroxide 0,5 mol*l 2.5l VWRC85520.320E
Sodium hydroxide 0,5 mol*l 25l VWRC85520.460E
Sodium hydroxide 2 mol*l 2.5l VWRC85521.320E
Sodium hydroxide 2 mol*l 25l VWRC85521.460E
Sodium hydroxide 6 mol*l 2.5l VWRC85522.320E
Sodium hydroxide 6 mol*l 25l VWRC85522.460E
Sodium hydroxide 25% 2.5l VWRC85523.320E
Sodium hydroxide 25% 25l VWRC85523.460E
Sodium hydroxide 50% 2.5l VWRC85524.320E
Sodium hydroxide 50% 25l VWRC85524.460E
Hydrochloric acid 1 mol 2.5l 85447.320E
Hydrochloric acid 1 mol 25l 85447.460E
Hydrochloric acid 2 mol 2.5l 85399.320E
Hydrochloric acid 2 mol 25l 85399.460E
Hydrochloric acid 5 mol 2.5l 85400.320E
Hydrochloric acid 5 mol 25l 85400.460E
Hydrochloric acid 6 mol 2.5l 85401.320E
Hydrochloric acid 6 mol 25l 85401.460E
Hydrochloric acid 10% Ph. Eur 10l 5594.9010E
Hydrochloric acid 10% Ph. Eur 210 kg 5594.9200E