Would you like to log in again?
VELP® UDK series Kjeldahl distillation units
Special Offer!
VELP® UDK series Kjeldahl distillation units
VALID UNTIL: 31/12/2017
Choose yout level of automation between models UDK 129 and UDK 139
Extremely reliable quantification of nitrogen/protein content with exclusive technologies and premium materials.
  • Patented titanium condenser for unmatched performance ensuring high performance and considerable water saving - the titanium condenser ensures that distillate temperature always remains below the safe threshold value of 35 °C
  • The patented steam generator offers unique performance and total safety (no internal pressure). The VELP steam generator is maintenance-free and has a built-in safety thermostat
  • The UDK 129 far exceeds expectations for an entry-level distillation unit - automatic NaOH addition
  • The semi-automatic Kjeldahl distillation unit UDK 139 runs automatically after setting distillation time, sodium hydroxide and water addition
  • 99,5% nitrogen recovery rate
Description Cat. No. Promo price Quantity
UDK 129 Kjeldahl distillation unit with patented steam generator and titanium condensers - LCD display 701-0224E
UDK 139 semi-automatic Kjeldahl distillation unit with patented steam generator and titanium condensers - 3.5" touch screen 701-0226E