Would you like to log in again?
VELP® flocculators
Great Offer!
VELP® flocculators
VALID UNTIL: 31/12/2017
Make your choice from VELP's range of jar tester for environmental tests
The perfect solutions for separation of pollutants in wastewater treatment plants.
  • JLT flocculators have four and six positions with common speed from 10 to 300 rpm
  • FC4S and FC6S jar testers have four and six positions independently controlled (10/15/30/45/60/90/120/150/200/300 rpm)
  • Stirring rods are adjustable and do not require any extra tools
  • FP4 is the portable solution for analysis on-site (20/40/50/100200 rpm)
  • The illuminated panels shows readout changes in the beakers
Model Stirring places Specifications Cat. No. Promo price Quantity
FC4S - independent position control 4 Illuminated back panel stirring rods adjustable in height without tools 705-1503E
FC6S - independent position control 6 Illuminated back panel stirring rods adjustable in height without tools 705-1504E
JLT 4 4 Timer for unattended operations & illuminated back panel 705-1501E
JLT 6 6 Timer for unattended operations & illuminated back panel 705-1502E
FP4 - portable jar tester 4 Battery power / car plug & selectable operating time 705-1500E