Would you like to log in again?
VELP® BOD EVO Sensor System
VELP® BOD EVO Sensor System
VALID UNTIL: 31/12/2017
Get BOD values from the sensor directly to your PC without opening the incubator and manage the data with BODSoft™
The wireless architecture allows fully automated remote system management, ensuring precise measurement and ease of use.
  • Remote operation with wireless technology
  • Wireless DataBox records transmitted data
  • Dedicated BODSoft software processes the data
  • Direct read-out in mg/l (ppm) and graph creation on PC
  • Measurement on scales of 90, 250, 600 and 999 ppm BOD
Description Cat. No. Promo price Quantity
BOD EVO Sensor System 6 (230 V) including Wireless Databox™, BODSoft™ and cable 705-0779E
BOD EVO Sensor System 6 (230 V) 705-0780E