Would you like to log in again?
Uvex pheos CX2 safety spectacles demonstrate excellent performance
Winner at work!
Uvex pheos CX2 safety spectacles demonstrate excellent performance
View video1
View video2
Suitable for most disciplines
The new uvex pheos CX2, takes uvex’s best selling spectacle to new heights. The X-tended eye shield and brow guard, with injection moulded, soft component technology, provides reliable protection against splash and particle ingress. The four-point frame check ensures the spectacle fits the wearer perfectly to deliver reliable protection. The distinctive design and shape of the side arms, together with the innovative X-Twist technology, allows the uvex pheos CX2 to adapt to individual head widths providing a comfortable, non slip fit. uvex pheos CX2 safety spectacles achieve outstanding performance for the protection in daily working life.
  • X-Twist technology
  • Scratch resistant and anti-fog
  • X-tended eye shield
  • Fold easily
  • Perfect ventilation
Anti-UV (5-2,5), lenses Colour Cat. No. Price Quantity
Grey PC White/black 111-2148E
Clear PC Black/blue 111-2149E
Clear PC Black/grey 111-2150E
Clear PC Red/grey 111-2151E
Clear PC Blue/grey 111-2332E