Would you like to log in again?
Urine Collection Systems
Urine Collection Systems
Urine analysis is one of the most common non-invasive methods for initial patient health screening. It is carried out in vast numbers in microbiology and clinical chemistry laboratories throughout the world.
VWR can offer you the consumables you need for automated processes and urine collection systems offering quick, simple and secure transfer of urine samples from collection containers to a primary analysis tube. The range of containers and tubes are perfect for microbial and microscopic sample analysis.
These systems offer a complete solution for laboratories looking to streamline urine collection and analysis.
  • Reduction of sample handling and contamination
  • Evacuated sample transfer eliminates spills and odour
  • Integrated transfer systems saving time and costs
  • Tubes designed specifically to fit automated urinalysis systems, including: Sysmex UF100, Sysmex UF1000i, Iris IQ 200, and Menarini sediMAX
Download Flyer