Would you like to log in again?
Tyvek® IsoClean® IC 183 - DS unhooded disposable overalls
NEW!
Tyvek® IsoClean® IC 183 - DS unhooded disposable overalls
Suitable for GMP Class A/B (ISO Class 5) cleanrooms
  • Gamma sterilised and clean processed to minimise particle shedding
  • Folded for aseptic donning and individually packed in an ISO Class 4 cleanroom
  • Bound seams and neck reduce particle release - the lowest in the DuPont portfolio
  • Double bagged (option DS) - inner and outer easy tear, validated, cleanroom bag to support risk management
  • Minimises the risk of cross contamination
  • Superior manufacturing and packaging quality
The ideal combination of protection, quality, comfort and contamination control for your peace of mind. Model IC183 B available with accessories (hood, boot covers, sleeves et al).
Description Pk Cat. No. Price Quantity
TYVEK® IsoClean® unhooded coverall, IC 183 B (option DS), sterile
Small 25 113-8628E
Medium 25 113-8629E
Large 25 113-8630E
X-Large 25 113-8631E
XX-Large 25 113-8632E
XXX-Large 25 113-8633E
TYVEK® IsoClean® frock with bound neckunhooded coverall, IC 270 B (option MS), sterile
Small 30 113-8634E
Medium 30 113-8635E
Large 30 113-8636E
X-Large 30 113-8637E
XX-Large 30 113-8638E
XXX-Large 30 113-8639E
TYVEK® IsoClean® boot cover, IC 458 B (option MS), sterile
Small 100 113-8640E
Medium 100 113-8641E
Large 100 113-8642E
X-Large 100 113-8643E
TYVEK® IsoClean® sleeve, IC 501 B (option MS), sterile
One size 100 113-8644E
TYVEK® IsoClean® hood with ties, IC 668 B (option MS), sterile
One size 100 113-8645E
TYVEK® IsoClean® bouffant, IC 729 WH (option MS), sterile
One size 250 113-8646E
TYVEK® IsoClean® hood & mask, IC 928 B (option MS), sterile
One size 100 113-8647E
Sierra™ mask, ML 7360 (option OS), sterile
One size 250 113-8648E