Would you like to log in again?
Antibody purification and analysis
White paper on antibody capture and analysis
Antibody purification and analysis
VALID UNTIL: 31/10/2017
Modern solutions for purification and QC of monoclonal antibodies.
Development and production of antibodies for therapeutic use require specific chromatographic tools.
  • High purity within one step
  • Fast analysis of aggregates and fragments
  • Robust and reliable methods
Description Pk Cat. No. Price Quantity
MiniChrom AF-rProtein A HC-650M, 5 ml polymer 8 mm ID 1 554-5787E
AF-rProtein A HC-650F, 45 µm; 1,000 Å polymer 1x10 ml 1 554-5788E
TSKgel SuperSW mAb HR (7,8x300 mm, 4 µm) 1 554-5613E
TSKgel SuperSW mAb HTP (4,6x150 mm, 4 µm) 1 554-5614E
TSKgel UltraSW Aggregate (7,8x300 mm, 3 µm) 1 554-5615E
TSKgel UP-SW3000 (4,6x300 mm, 2 µm) 1 554-5317E
TSKgel UP-SW3000 (4,6x150 mm, 2 µm) 1 554-5318E