Would you like to log in again?
TOMO® adhesion slides
TOMO® adhesion slides
VALID UNTIL: 31/10/2017
Recommended for use on Ventana Benchmark staining and automated IHC platforms.
  • High adhesion to reduce tissue loss and decrease the number of repeated tests
  • Background staining reduced in special dyes
  • Frosted ends optimised for thermal slide printers; clear and consistent printing
  • High flatness
  • Reduced surface leaching, increased stabilility and longevity of slides
Colour LxW Pk Cat. No. Promo price Quantity
White 72x25 mm 100 631-1109E
Yellow 72x25 mm 100 631-1128E
Pink 72x25 mm 100 631-1129E
Blue 72x25 mm 100 631-1132E
GrEn 72x25 mm 100 631-1133E