Would you like to log in again?
Thermo Scientific™ Nunc Plates Selection Guide
Thermo Scientific™ Nunc Plates Selection Guide
Quality and innovation for your cell culture, cell-based assay, screening and immunoassay applications
Thermo Scientific™ has over 50 years of experience in designing and manufacturing plates for researchers’ increasingly sophisticated applications. The result is a very wide offering of the highest quality microplates for cell-based assays, cell-imaging, screening or immunoassays. Common to all plates: adherence to ISO 13485 quality standards, easy availability of product certification and quick access to knowledgeable technical support personnel.
Thermo Scientific continues to innovate with the newly launched Thermo Scientific™ Nunc™ Edge 2.0 96-well plate uniquely designed with a surrounding moat that can be filled by sterile liquid or gel to prevent evaporation and preserve cell viability in the 96 wells of the plate during cell‑based assays.
Download the selection guide to find the best plate suited to your application.