Would you like to log in again?
Stainless steel furniture for cleanrooms
Now available on web!
Stainless steel furniture for cleanrooms
Discover this new broad range of cleanroom furniture
The basic principle that guides the process of designing elements is to provide ease of use and maintenance of furniture. Tinman considers all possible aspects of the future use of our furniture in cleanrooms in order to optimise working conditions for each user by offering them the highest level of use and comfort.
  • Closed circuit welded joints
  • All rounded corners
  • Minimal joints
  • Solid or perforated
  • Modular furniture design
Are you planning to equip your cleanroom? VWR offers you a new range of high products are made from AISI 304 and optional AISI 3016 - high gloss polished. Made from circular elements, easy to clean and simple to maintain. 
Description Dimensions Material Cleanroom suitability Cat. No. Price Quantity
Desk, round front edge, adjustable levelling feet, double stainless steel sheet 600x600x750 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1347E
Desk, round front edge, adjustable levelling feet, double stainless steel sheet 800x600x750 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1348E
Desk, round front edge, adjustable levelling feet, double stainless steel sheet 1000x600x750 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1349E
Desk, round front edge, adjustable levelling feet, 40% perforation, hidden reinforcement 600x600x900 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1350E
Desk, round front edge, adjustable levelling feet, 40% perforation, hidden reinforcement 800x600x900 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1351E
Desk, round front edge, adjustable levelling feet, 40% perforation, hidden reinforcement 1000x600x900 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1352E
Shelf, height adjustable, storing, rounded edges 766x400x1800 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1353E
Shelf, height adjustable, storing, rounded edges 966x400x1800 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1354E
Shelf, height adjustable, storing, rounded edges 1166x400x1800 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1355E
Bench, step over, rounded edges, modular, double stainless steel sheet 600x350x450 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1356E
Bench, step over, rounded edges, modular, double stainless steel sheet 800x350x450 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1357E
Bench, step over, rounded edges, modular, double stainless steel sheet 1000x350x450 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1358E
Garment rack, hangers (fixed), modular with perforated frame 530x500x1800 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1359E
Garment rack, hangers (fixed), modular with perforated frame 930x500x1800 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1360E
Transport cart with adjustable shelves and castors 990x525x1000 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1361E
Transport cart with adjustable shelves and castors 1090x525x1000 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1362E
Step stool, one step, perforated with levelling feet and treads 420x720x440 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1363E
Dispenser, flat top, disposable gloves, bevelled exit, swing open lid and viewing slots 204x211x590 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1364E
Dispenser, wall mounted spectacles and goggles, acrylic 300x200x400 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1365E
Waste bin, plastic bag, holder and castors 400x400x1000 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1366E
Mirror, wall mounted, 8R polished with bevelled edge 490x1180x255 mm AISI 304 /2R – high gloss polish ISO Class 4 /GMP Class A 132-1367E