Would you like to log in again?
VWR will be introducing a new supplier for stainless steel cleanroom equipment
VWR will be introducing a new supplier for stainless steel cleanroom equipment
Excellent cleaning accessibility and design
    Are you planning to equip your cleanroom? From early 2017 VWR will offer you a new range of high quality cleanroom furniture.
  • Construction material: AISI 304 and optional AISI 3016 - high gloss polish
  • Products are made from circular elements, easy for cleaning and very simple to maintain
  • Furniture is made solid or perforated - perforations enable air permeability of 40%, for unobstructed laminar airflow
  • Very good resistance to chemicals and all regular cleanroom disinfectants (alcohols, biocides and sporocides)
  • Full range available soon at  www.vwr.com/cleanroom