Would you like to log in again?
Thermo Scientific Varioskan LUX Microplate Multimode Reader
Thermo Scientific Varioskan LUX Microplate Multimode Reader
Designed for fast and reliable results even for the most challenging applications including cell based assays
  • Five reading technologies: Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen / AlphaLISA, and Time-Resolved Fluorescence
  • Autocalibration and self diagnostics for reliable performance day to day, run to run
  • SMART safety features quarantee failsafe operation
  • SkanIt Software enables easy to use, effortless analysis
  • Modular and upgradable design enables to meet current and future needs
  • Integrated gas module allow optimal conditions for cell based assays with atmospheric control of C02 and O2
Long-term cell-based assays in a microplate reader require maintenance of consistent physiological conditions for the cultured cells inside the instrument for up to several days. In this study we demonstrate that cells incubated in the Varioskan LUX instrument with an integrated gas module survive and proliferate well.