Would you like to log in again?
Thermo Scientific™ Finnpipette™ F2 manual pipettes
Thermo Scientific™ Finnpipette™ F2 manual pipettes
VALID UNTIL: 31/12/2017
Upgrade any old pipette to Thermo Scientific's Finnpipette F2 manual pipettes and get 35% off!
Upgrade to a pipette that’s rugged and built to excel.
  • RUGGED AND DURABLE: The Finnpipette F2 is made of tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light
  • FULLY AUTOCLAVABLE: Decontamination of the Finnpipette F2 is simple because there’s no need to take apart/re-assemble the F2 for autoclaving, minimising disruption and downtime
  • MAINTENANCE IS EASY: Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave
Finnpipette F2 rugged construction and ease of maintenance for your more demanding applications.
Description Cat. No. Promo price Quantity
Single channels
Finnpipette F2, 0,2 - 2 μl, micro 613-2696E
Finnpipette F2, 0,5 - 5 μl, micro 613-2697E
Finnpipette F2, 1 - 10 μl, micro 613-2698E
Finnpipette F2, 1 - 10 μl 613-2699E
Finnpipette F2, 2 - 20 μl, micro 613-2700E
Finnpipette F2, 2 - 20 μl 613-2701E
Finnpipette F2, 5 - 50 μl, micro 613-1486E
Finnpipette F2, 5 - 50 μl 613-1487E
Finnpipette F2, 10 - 100 μl 613-2702E
Finnpipette F2, 20 - 200 μl 613-2703E
Finnpipette F2, 100 - 1000 μl 613-2704E
Finnpipette F2, 0,5 - 5 ml 613-2705E
Finnpipette F2, 1 - 10 ml 613-2706E
Multi channels
Finnpipette F2, 8-channel, 1 - 10 μl 613-2720E
Finnpipette F2, 8-channel, 5 - 50 μl 613-2721E
Finnpipette F2, 8-channel, 10 - 100 μl 613-2722E
Finnpipette F2, 8-channel, 30 - 300 μl 613-2723E
Finnpipette F2, 12-channel, 1 - 10 μl 613-2724E
Finnpipette F2, 12-channel, 5 - 50 μl 613-2725E
Finnpipette F2, 12-channel, 10 - 100 μl 613-2726E
Finnpipette F2, 12-channel, 30 - 300 μl 613-2727E
Finnpipette F2, 16-channel, 1 - 10 μl 613-2728E
Finnpipette F2, 16-channel, 5 - 50 μl 613-2729E