Would you like to log in again?
Thermo Scientific™ MabPac RP HPLC columns, reversed phase columns for characterising biotherapeutics
Now with 25% off!
Thermo Scientific™ MabPac RP HPLC columns, reversed phase columns for characterising biotherapeutics
VALID UNTIL: 30/04/2018
Get optimised reversed-phase LC-MS separations with columns designed for the job
Try this HPLC column, designed for mAb characterisation, including the separation of mAb and variants.
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency columns with lower carry-over than silica-based columns
  • Excellent MS compatibility
  • Wider operating pH range (0 - 14) and high temperature stability (110 °C)
  • Fast separation capability for high throughput separations
For more information  
  Desgnation Cat. No. Promo price Quantity
  MAbPac RP, Analytical 4 µm, 1.0 x 50 mm 554-5939P  
  MAbPac RP, Analytical 4 µm, 1.0 x 150 mm 554-5940P  
  MAbPac RP, Analytical 4 µm, 1.0 x 100 mm 554-5941P  
  MAbPac RP, Analytical 4 µm, 3.0 x 100 mm 554-4732P  
  MAbPac RP, Analytical 4 µm, 3.0 x 50 mm 554-4733P  
  MAbPac RP, Analytical 4 µm, 2.1 x 100 mm 554-4735P  
  MAbPac RP, Analytical 4 µm, 2.1 x 50 mm 554-4736P  
  MAbPac RP, Guard Cartridges 4 µm, 3.0 x 10 mm, 2/pk 554-4734P  
  MAbPac RP, Guard Cartridges 4 µm, 2.1 x 10 mm, 2/pk 554-4737P  
  V-2 guard holder SST 554-4528P