Would you like to log in again?
Nunc™ plate selection guide for cell-based assays - screening - immunoassays
Learn more
Nunc™ plate selection guide for cell-based assays - screening - immunoassays
VALID UNTIL: 31/10/2017
Nunc™ plate selection guide for cell-based assays - screening - immunoassays
Select in 3 easy steps the best Nunc plate for your application.
  • Quality and innovation for your assays
  • Thermo Scientifichas over 50 years of experience in designing and manufacturing plates for researcher's increasingly sophisticated applications. The result is a very wide range of the highest quality microplates for cell-based assays, cell imaging, screening or immunoassays. Common to all plates: Adherence to ISO 13485 quality standards, easy availability of product certification, and quick access to knowledgeable technical support personnel
  • Thermo Scientific continues to innovate with the newly launched Thermo Scientific Nunc Edge 2.0 96-well plate, uniquely designed with a surrounding moat that can be filled with sterile liquid, or gel, to prevent evaporation and preserve cell viability during cell‑based assays
  • Download the selection guide to find the best plate suited to your application