Would you like to log in again?
Thermo Scientific Heratherm™ ovens
Thermo Scientific Heratherm™ ovens
VALID UNTIL: 31/08/2017
Large capacity ovens for high sample volumes
Natural convection ovens for gentle heating and drying.
  • Simple, microprocessor-based, touch button controls
  • 180° door opening for convenient access
  • Automatic over-temperature alarm system
  • Built-in timer
  • Lockable castors for easy mobility and stability
Model OGS400 OGS750    OGS750-3P
Capacity (l) 419 774 774
Temperature range (°C)  Ambient +10...250 Ambient +10...250 Ambient +10...300
Temperature variation (spatial) at 37 °C  (°C)  ±3,0  ±3,7  ±3,7
Temperature fluctuation (time) at 37 °C  (°C)  ±0,5  ±0,5  ±0,5
Shelves supplied (max.) 2 (39) 2 (39) 2 (39)
Max. load per shelf (kg) 40 40 40
Int. W×D×H (mm) 544x590x1307 1004x590x1307 1004x590x1307
Ext. W×D×H (mm) 778x770x1545 1261x770x1545 1261x770x1545
Power consumption (W) 2400 3000 6350
Weight (kg) 136 182 182
Description Cat. No. Promo price Quantity
General Protocol oven OGS 400, 230 V 466-0377E
General Protocol oven OGS 750, 230 V 466-0379E
General Protocol oven OGS 750-3P, 400 V, 3 phase 466-0380E
Additional wire shelf for 400 l models 466-0446E
Additional wire shelf for 750 l models 466-0447E