Would you like to log in again?
ProPac™ WCX-10 Premier Bio-LC columns
ProPac™ WCX-10 Premier Bio-LC columns
VALID UNTIL: 30/10/2017
The industry standard for high resolution and high efficiency analysis of MAbs, other proteins and associated variants.
  • Ideal for characterisation studies and QA/QC assessment
  • Unmatched column-to-column and lot-to-lot reproducibility
  • Resolve isoforms that differ by a single charged residue
  • A thin, hydrophilic layer grafted to the particle core eliminates unwanted secondary interactions
  • Based on stationary phases composed of 10 µm non porous polymeric beads
ProPac WCX-10 guard columns
I-Ø×L Pk Cat. No. Price Quantity
2,0x50 mm 1 554-5304E
4,0x50 mm 1 554-5305E
ProPac WCX-10 columns
Column format I-Ø×L Pk Cat. No. Price Quantity
Analytical 4,0x50 mm 1 554-5300E
Analytical 4,0x100 mm 1 555-7453E
Analytical 4,0x150 mm 1 555-7454E
Analytical 4,0x250 mm 1 554-5301E
Analytical 9,0x250 mm 1 554-5302E
Analytical 22,0x250 mm 1 554-5303E
Semi-preparative 2,0x250 mm 1 555-7441E