Would you like to log in again?
DNAPac™ RP Columns
Launched in 2016
DNAPac™ RP Columns
High Performance Reversed-Phase LC Columns for superior oligonucleotides separations
High resolution and high throughput
  • Highly stable to extremes of pH (0-14), and temperature (5-110°C)
  • Operates with mobile phases compatible with direct mass spectrometry
  • Compatible with both HPLC and UHPLC instruments
  • Resolves both small and very large dsDNA segments
  • Analysis of double-stranded (ds) DNA/RNA fragments
Learn more by reading the application note below. 
Description Cat. No. Price Quantity
DNAPac RP, Analytical, 4 µm, 3.0 x 100 mm HYPE088919
DNAPac RP, Analytical, 4 µm, 3.0 x 50 mm HYPE088920
DNAPac RP, Guard Cart, 4 µm, 3.0 x 10 mm (2/pk) HYPE088921
DNAPac RP, Analytical, 4 µm, 2.1 x 100 mm HYPE088923
DNAPac RP, Analytical, 4 µm, 2.1 x 50 mm HYPE088924
DNAPac RP, Guard Cart, 4 µm, 2.1 x 10 mm (2/pk) HYPE088925