Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
Eutech™ EXPERT pH and conductivity pocket testers
Eutech™ EXPERT pH and conductivity pocket testers
VALID UNTIL: 31/12/2017
Save money and make your testing simpler
Accurate 'on the go' solutions for water analysis.
  • Easy to operate and intuitive user interface with one-push calibration
  • Large four-digit, double-line display with simultaneous temperature display
  • Automatic temperature compensation: Adjusts readings according to temperature change automatically for greater accuracy
  • Three-point auto calibration with auto buffer recognition (EXPERT pH) allows the tester to measure both accurately and efficiently
  • Durable keypad and IP 67 waterproof – lightweight tester floats on water for easy retrieval
Description Cat. No. Promo price Quantity
Waterproof pH tester, Eutech Expert pH 663-0308E
Waterproof conductivity tester, Eutech Expert CTS 663-0307E