Would you like to log in again?
Sterile IPASEPT® Premium
High quality IPA!
Sterile IPASEPT® Premium
Produced in our own cleanrooms!
  • Produced with USP WFI water
  • Supplied in bag-in-bottle packaging, a trigger spray preventing air going into the bottle
  • The whole production process is tightly monitored by our QA department
So from the raw material, via the filling environment, the process itself, the packaging technology until the final release, we can give full quality assurance. The product also comes with a full validation package.
Description Pk Cat. No. Promo price Quantity
IPASEPT® 70 Premium, sterile, spray bottle, filtered, irradiated (>25 kGy) 1 l 94018.1006E