Would you like to log in again?
TitroLine® 7800 - the automatic universal titrator
TitroLine® 7800 - the automatic universal titrator
VALID UNTIL: 31/12/2017
High measuring accuracy raises confidence in results to a whole new level
The TitroLine® 7800 comes with additional IDS® measurement input which can perform pH, acidity, ORP, conductivity, salt, dissolved oxygen and volumetric Karl Fischer titrations.
  • Comes with many standard methods for common potentiometric and KF titration
  • In just one sample: Measurement of pH and conductivity plus titration of acid capacity
  • Karl Fischer titration – the method for determining water content
  • Measurement and calibration with the highest accuracy
  • Second digital measuring port for intelligent digital sensors (IDS®)
Description Includes Cat. No. Promo price Quantity
TitroLine® 7800 basic unit with magnetic stirrer TM 235 2 measuring inputs, 1 analogue and 1 digital (IDS) measuring input 661-0240E
Description Includes Cat. No. Promo price Quantity
Exchangeable unit, WA 20, 20 ml Integrated chip for reagent data, brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tip 661-0126E
pH combination electrode with sensor recognition Glass shaft, PT junction, electrolyte KCl 3 mol/l, Silamid® reference, temperature sensor PT1000, sphere membrane, A glass, 2 m fixed cable with DIN + 4 mm banana plug and recognition, length 120 mm, 12 mm Ø, -5...+100 °C, 0...14 pH 661-0243E
Conductivity cell with sensor recognition IDS® 4 pole cond. cell for small sample volumes, plastic shaft, 3 m cable with digital plug, sensor material graphite, cell constant 0,47 cm - 1, temp. sensor NTC 30 kOhm, min. immmersion depth 16 mm, length 120 mm, 15,3 mm Ø, -5...+80 °C 661-0272E