Would you like to log in again?
Micro pH electrodes
Micro pH electrodes
VALID UNTIL: 31/10/2017
For small sample volumes in life science and pharmaceutical applications
Measurement often takes place in small receptacles such as vials, small vessels, NMR tubes etc. Due to small sample volumes it is important to take care of the electrode's minimum immersion length in order to place the junction into the sample as well.
  • Reliable, reproducible results
  • Highest quality and long lasting
  • Accurate and highly precise
  • Suitable temperature range: −5 to +100 °C
Description Type ØxL Cat. No. Promo price Quantity
Glass shaft, platinum junction, electrolyte KCl 3 mol/l, Ag/AgCl reference system, spear membrane, A-glass, DIN plug, tapper NS 7.5, -5...+100 °C, 0...14 pH N 5800 A 5x96 mm 662-1267E
Same as N 5800 A, but with BNC plug N 5800 BNC 5x96 mm 662-1675E
Glass shaft, platinum junction, electrolyte KCl 3 mol/l, Ag/AgCl reference system, cylinder membrane, A-glass, DIN plug, tapper NS 7.5, -5...+100 °C, 0...14 pH N 6000 A 3x96 mm 662-6109E
Same as N 6000 A, but with BNC plug N 6000 BNC 3x96 mm 662-1676E
Glass shaft, platinum junction, electrolyte KCl 3 mol/l, Ag/AgCl reference system, cylinder membrane, A-glass, 1 m fixed cable with BNC-plug and recognition, tapper NS 7.5, -5...+100 °C, 0...14 pH N 6000 1M-BNC-ID 3x96 mm 662-1681E
Glass shaft, platinum junction, electrolyte KCl 3 mol/l, Ag/AgCl reference system, cylinder membrane, A-glass, 1 m fixed cable with DIN plug and recognition, tapper NS 7.5, -5...+100 °C, 0...14 pH N 6000 1M-DIN-ID 3x96 mm 662-1680E
Glass shaft, ceramic junction, electrolyte KCl 3 mol/l, Ag/AgCl reference system, cylinder membrane, A-glass, plug head, -5...+100 °C, 0...14 pH - suitable for NMR tubes N 6003 3x180 mm 662-1673E
Glass shaft, PT junction, electrolyte KCl 3 mol/l, Silamid® reference, temperature sensor PT1000, sphere membrane, A glass, 1 m fixed cable with BNC + 4 mm banana plug and recognition, -5...+100 °C, 0...14 pH A 157 1M-BNC-ID 12/5x40/130 mm 662-1679E
Glass shaft, PT junction, electrolyte KCl 3 mol/l, Silamid® reference, temperature sensor PT1000, sphere membrane, A glass, 1 m fixed cable with DIN + 4 mm banana plug and recognition, -5...+100 °C, 0...14 pH A 157 1M-DIN-ID 12/5x40/130 mm 662-1678E