Would you like to log in again?
Safety funnel with level control
Keep filling levels, in non transparent containers, safe and under control
Safety funnel with level control
VALID UNTIL: 30/11/2017
World’s first funnel with level control for disposing of HPLC liquid solvent waste safe and easy.
  • 9,75 litres throughput per minute
  • Electrostatically conductive PE-HD
  • Grounding cable and clamp included
  • Removable sieve
  • Suitable for every safety cabinet