Would you like to log in again?
Quintix® - the benchmark standard balance for your laboratory
Limited time special offer!
Quintix® - the benchmark standard balance for your laboratory
VALID UNTIL: 30/09/2017
Effective features make your work flows easier
Intuitive touch screen, in combination with the most precise weighing technology, to achieve the best results.
  • Entirely new touch screen user-interface with built-in application programs
  • Fully automatic internal adjustment
  • Easier record-keeping using the balance's directly integrated PC functions
  • Plug & work connectivity to a Sartorius lab printer
  • Space saving footprint and easy to clean design
Model Readability Linearity (± mg) Weighing cap. (g) Cat. No. Promo price Quantity
Analytical semi-micro balances
QUINTIX35-1S 0,01 mg 0,1 30 611-3291E
QUINTIX65-1S 0,01 mg 0,1 60 611-3292E
QUINTIX125D-1S 0,01 / 0,1 mg 0,1 / 0,2 60 / 120 611-3290E
Analytical standard balances
QUINTIX64-1S 0,01 mg 0,2 60 611-3124E
QUINTIX124-1S 0,01 mg 0,2 120 611-2648E
QUINTIX224-1S 0,01 mg 0,2 220 611-2645E
Precision standard balances
QUINTIX213-1S 0,001 mg 2 210 611-2646E
QUINTIX313-1S 0,001 mg 2 310 611-2643E
QUINTIX513-1S 0,001 mg 2 510 611-2639E
QUINTIX613-1S 0,001 mg 2 610 611-3125E
QUINTIX412-1S 0,01 mg 30 410 611-3127E
QUINTIX612-1S 0,01 mg 30 610 611-2638E
QUINTIX1102-1S 0,01 mg 30 1100 611-2649E
QUINTIX2102-1S 0,01 mg 30 2100 611-2647E
QUINTIX3102-1S 0,01 mg 20 3100 611-2644E
QUINTIX5102-1S 0,01 mg 20 5100 611-2640E
QUINTIX6102-1S 0,01 mg 20 6100 611-3130E
QUINTIX2101-1S 0,1 mg 300 2100 611-3126E
QUINTIX5101-1S 0,1 mg 300 5100 611-2641E
QUINTIX6101-1S 0,1 mg 300 6100 611-3128E
QUINTIX5100-1S 1,0 g 1000 5100 611-2642E
QUINTIX6100-1S 1,0 g 1000 6100 611-3129E