News
RightCycle* - the first true recycling programme for Kimtech* nitrile gloves and garments
What’s the cost of sustainability?
From now on your used, uncontaminated Kimtech* nitrile gloves and garments are no longer waste but can be recycled with the RightCycle* Program from Kimberly-Clark Professional*
SecureFit™ 600 protective eyewear for better vision and comfort
NEW!
Get comfortable with changes!
Chemical glove selection simplified
Personalised chemical glove assessment
Ansell Chemical Guardian™ simplifies the chemical glove selection process for your unique set of chemicals
Uvex pheos CX2 safety spectacles demonstrate excellent performance
Winner at work!
Suitable for most disciplines
The new VWR Industrial Safety Solutions catalogue has arrived!
Products and solutions on more than 400 pages
Request your free copy or easily download the PDF
Microgard® protective clothing - featuring 2000Ts Plus
Product of choice for pharmaceutical workers around the world!
MICROGARD® 2000 Ts Plus is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0.01 microns in size*. (*EMSL test method).
Microflex® 93-260 nitrile and neoprene double-coated disposable gloves
Thinnest chemical resistant synthetic composite disposable glove
VWR® CHF5CB coveralls
Don´t compromise on comfort or safety
Absorbents (mats, rolls and SOC's) for cleaning up oil, water and chemical-based spills
NEW!
VWR absorbents and spill containment products help you keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
Lockout Tagout (LoTo)
Protects workers from risks posed by live machinery or electricity
VWR presents 3M solutions to solve hearing protection challenges
NEW!
Get hearing protection right - on site and in sight
EN374-3:2003 is to be replaced by EN16523-1 by the end of the year
Certainty at first sight!
Both methods are technically very similar, but there will be significant changes in requirements. The list of challenge chemicals will increase from 12 to 18 and the Beaker symbol will be replaced by 3 categories of Erlenmeyer flask pictogram.
Holistic concepts for personal protective clothing and workwear
Custom-made solutions for workplaces and employees!
Thorough understanding of customer's individual requirements for innovative and creative solutions.
Your health and safety is our concern
Contact us for training and solutions for your protection!
VWR provides professional advice in selecting the appropriate safety products for your lab environment.
VWR presents educational material on industrial and lab safety solutions!
Our aim is to support you beyond our comprehensive portfolio of safety products. Go to VWR.com/safety to review the extensive educational material on safety, protection and hygiene topics. Just some examples of personal protection.
VWR® Protection Solutions
VWR is your partner for industrial safety supplies & services
VWR presents 3M solutions to solve hearing protection challenges
NEW
3M is passionate about protecting the hearing of workers in all types of environments.
BBP™ 35-37 multicolour sign and label printers
Because safety means the world to you
BBP™12 label printer laboratory Identification
Identify every sample with the BBP12 label printer!
Industrial safety and protection solutions
NEW!
The complete overview at a glance!