Would you like to log in again?
Merck
Merck
Merck analytical reagents are not only specified in accordance with the American ACS Standard but also with Reag.Ph Eur. Therefore these reagents comply with the reagents part of both the American Pharmacopoeia and the European Pharmacopoeia.
The reliability of Merck’s analytical reagents gives you multiple benefits:
  • Compliance with the ACS standard and Reag. Ph Eur
  • Rapid updating of new specification requirements
  • Validated analytical methods
  • Comprehensive range of analytical reagents
  • Lower costs due to reliable audits
  • Lower validation effort due to internationally consistent product quality
The validation plan for ACS reagents has been carried out according to the very strict validation instructions for pharmaceutical starting materials (e.g. according to USP and ICH Q2A ‘Guidelines for Industry , Text on Validation of Analytical Procedures’)
Just type the substance name into the search box and refine the display to just Merck. The description will show you the product you need.
Just type the substance name into the search box and refine the display to just Merck. The description will show you the product you need.