Would you like to log in again?
Moisture Analysis
Moisture Analysis
VWR offers a choice of infrared or halogen heating, ceramic or coiled quartz element moisture analysers for routine and precision measurements.
  • Infrared heating - no glass (MB23), halogen for fast results (MB25), heating elements with glass or aluminium panels (MA35)
  • High precision moisture analysers: Halogen (MB35, MB45, MA 100), ceramic or coiled quartz elements (MA100 and MA150)
  • A range of verified versions (OCE) for use in legal metrology is available