Would you like to log in again?
VWR Chemicals
VWR Chemicals
AnalaR NORMAPUR high purity analytical grade reagents combine the best quality with competitive prices.
This comprehensive product range offers over 400 reagents suitable for a multitude of analytical applications. Large choice of packaging and pack sizes: in glass, plastic or plastic-coated glass bottles. Larger pack sizes, up to 200 l, are also available for many lines. All BDH Prolabo products show an expiry date on the product label and certificate of analysis. Each batch of reagents is tested against an exacting specification to ensure a consistent performance time after time. The specifications quote guaranteed maximum permitted limits of impurity, not ‘typical analyses’. Certificates of analysis are easily obtained from our web site.
Simply enter the chemical you're looking for into the search engine to see what is available.
Please contact VWR if you need a large volume and the pack size you require is not listed.