Would you like to log in again?
Evaporation
Evaporation
Rotary Evaporators
Used for the efficient and gentle removal of solvents from samples by evaporation, under reduced atmospheric pressure, thus allowing the solvents to evaporate faster at low temperatures.
Physical or thermal separation is a common technique in many chemistry laboratories. VWR has an extensive range of equipment for automating these techniques, including the great value for money joint IKA/VWR Collection rotary evaporators, plus all the glassware you need to create your own combinations.