Would you like to log in again?
MicroRNA profiling - confidence in your results and flexibility in assay design
Test now for FREE!
MicroRNA profiling - confidence in your results and flexibility in assay design
VALID UNTIL: 31/10/2017
The qScript microRNA system uses a powerful 2-step RT-qPCR reaction suitable for miRNA detection from a variety of samples.
  • Open technology provides flexibility to design your own assay primers
  • Compatible with both total RNA- and microRNA-enriched samples
  • Detects rare miRNAs - linear assay performance over a broad range of input RNA
  • Seamlessly profile hundreds of miRs from a single RT reaction
  • Eliminates inefficient stem-loop priming and costly hydrolysis probes
Description Pk Cat. No. Price Quantity
PerfeCTa SYBR Green SuperMix 100 R 733-2591E
qScript MicroRNA cDNA Synthesis kit 25 R 733-2218E