Would you like to log in again?
KF Titration
KF Titration
Choice of compact volumetric or coulometric Karl Fisher titrators and related reagents for water determination.
  • Volumetric Karl Fischer titrators are for medium to large volumes of water (mg range) combining fast and precise water content determination with easy operation
  • Coulometric Karl Fischer titrators are for small water volumes (µg range) combining fast and precise water content determination with very simple operation
  • Karl Fischer reagents