Would you like to log in again?
Cleaning Solutions
Cleaning Solutions
Sodium and potassium hydroxide solutions are routinely used for post-production cleaning.
VWR stock standard 50%, 40%, 35%, 32% and 30% solutions ready-to-use and in various grades. In addition we can supply a custom product tested to pharmacopoeia levels if needed.
We can also offer a range of isopropanol based cleaning solutions for use in cleanrooms. They are available with spray nozzles and can be double wrapped and irradiated if needed. Ask for a custom format if required.