Would you like to log in again?
Nanosep Centrifugal Devices
Order your free sample pack now!
Nanosep Centrifugal Devices
Ideal for numerous applications including the concentration, purification, and desalting of oligonucleotides, DNA, RNA and proteins
  • Ensures rapid processing of samples from 50 to 500 µl
  • Isolate DNA from agarose gel slices
  • Separate oligonucleotides and RNA from acrylamide gels
  • Clean up labeling and PCR reactions
Gel electrophoresis separates DNA molecules at a very high resolution based on size. However, the majority of scientists need to do much more than simply analyze restriction digest DNA fragments on agarose gels. For these applications, DNA molecules are cut, ligated, sequenced, transformed, electrophoresed, and labeled.
In addition, desalting, buffer exchange, or elution from agarose may be required during any of a number of procedures. Ultrafiltration allows the rapid purification and concentration of the DNA using Nanosep devices. With this method there is very little handling, yields are high, DNA binding is low, DNA is undamaged, and the concentrated DNA is free of contaminants and ready for further reactions. Nanosep devices are available in a wide range of MWCOs and are color-coded for easy identification.
Description Cat. No. Quantity
Nanosep MF Centrifugal Devices with Bio-Inert® Membrane 0.45 μm PALLSODM45C33
Nanosep MF Centrifugal Devices with Bio-Inert® Membrane 0.2 μm PALLSODM02C33
Nanosep Centrifugal Devices with Omega Membrane 300K PALLSOD300C33
Nanosep Centrifugal Devices with Omega Membrane 100K PALLSOD100C33
Nanosep Centrifugal Devices with Omega Membrane 30K PALLSOD030C33
Nanosep Centrifugal Devices with Omega Membrane 10K PALLSOD010C33
Nanosep Centrifugal Devices with Omega Membrane 3K PALLSOD003C33