Would you like to log in again?
Try Before You Buy - Mag-Bind® Normalizer Kit
FREE Sample
Try Before You Buy - Mag-Bind® Normalizer Kit
  • Automation-friendly format
  • Isolate PCR or plasmid DNA
  • Adjust magnetic bead or elution buffer to achieve desired concentration
The Mag-Bind ® Normalizer Kit allows simple, high throughput purification and normalisation of PCR and plasmid product concentration via limited binding magnetic beads. Receiving samples from multiple sources causes multiple challenges for high throughput workflow including the various quality and quantity of DNA received. To achieve optimal results with many downstream applications the DNA concentrations must be within a optimal range. The Mag-Bind ® Normalizer Kit allows for various input amounts of DNA to be added and the final elute contains a constant DNA concentration. During the process the samples are bound, washed and eluted providing a consistent DNA quality product. The Mag-Bind ® Normalizer Beads are designed for use with PCR products or plasmid DNA.