Would you like to log in again?
OHAUS® MB23 and MB25 moisture analysers
OHAUS® MB23 and MB25 moisture analysers
VALID UNTIL: 31/12/2017
Speed, simplicity and affordability for routine moisture analysis
High quality construction with a compact design offering dependable, accurate results.
  • Infrared (MB23) or halogen (MB25) heating with 0,1% or 0,05% readability
  • Compact, easy to clean design ideal for frequent use with a wide range of samples
  • Bright backlit LCD display and easy one-button set up and menu navigation
  • Choice of displayed results as percent moisture, percent solids or weight (g) with temperature and time
  • Robust ABS housing, metal pan support and metal pan handler with in-use cover as standard
Model Heating element Capacity x readability Temperature range (°C) Drying time Cat. No. Promo price Quantity
MB23 Infrared 110x0,01 g / 0,1% 50...160 in 5 °C increments Manual, automatic or timed 611-3869E
MB25 Halogen 110x0,005 g / 0,05% 50...160 in 5 °C increments Manual, automatic or timed 611-3870E