Would you like to log in again?
For a limited time, buy the SpectraMax M3 microplate reader for the price of the M2
Don’t miss this incredible offer on microplate readers!
For a limited time, buy the SpectraMax M3 microplate reader for the price of the M2
VALID UNTIL: 31/12/2017
Get more microplate reader functionality for a lower price!
The SpectraMax M series (M2, M2e, M3, M4, M5 and M5e) microplate readers all offer UV/Vis absorbance and fluorescence intensity measurements on microplates and cuvettes. So choose the model with the best combination of features for your lab.
  • As well as absorbance and fluorescence, you’ll get an additional luminescence mode with the M3 compared to the M2
  • Measure fluorescence readings from the top and bottom (ideal for cell-based assays)
  • The SpectraMax M3 is upgradable to the M4, M5 and M5e whereas the M2 is not
  • All this, plus savings of more than £6,000!
  • Free application note
Description Cat. No. Promo price Quantity
SpectraMax® M3 multi-mode microplate reader 736-0910E