Would you like to log in again?
Microgard® protective clothing - featuring 2000Ts Plus
Product of choice for pharmaceutical workers around the world!
Microgard® protective clothing - featuring 2000Ts Plus
MICROGARD® 2000 Ts Plus is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0.01 microns in size*. (*EMSL test method).
  • Anti-static  (according to EN 1149-5)
  • Low linting: Reduced risk of contamination
  • Silicone-free: Critical in spray painting applications
  • Comfort: Moisture vapour permeable to help reduce the risk of heat stress
  • Thumb loops help prevent sleeve movement when working above your head
  • Chin strap helps to reduce risk of cross-contamination
Acquired by Ansell in 2015, Microgard Limited is a leading manufacturer of chemical protective clothing and have introduced new technologies and designs to the market to improve wearer protection and comfort for the past 30 years.
MICROGARD® 2000 Ts Plus coverall, white
Size Pk Cat. No. Price Quantity
Small 45 113-8553E
Medium 45 113-8554E
Large 45 113-8555E
X-Large 45 113-8556E
XX-Large 45 113-8557E
XXX-Large 45 113-8558E