Would you like to log in again?
Contact Plates & Dipslides
Contact Plates & Dipslides
Contact plates available in blister, double or triple wrapped packaging complete with dipslides, all according to regulations. Available with TLHTh (tween, lecithin, histidine, thiosulphate) neutraliser accompanied by a certificate of irradiation.
Settle plates are irradiated for active or passive air monitoring. They come in blister or classical packaging with dipslides, all according to regulations.