Would you like to log in again?
Merck Millipore SupraSolv® solvents for headspace gas chromatography
Merck Millipore SupraSolv® solvents for headspace gas chromatography
VALID UNTIL: 31/10/2017
For the determination of residual solvents in drugs and active substances, according to ICH, Ph. Eur. and USP.
  • Specially designed for the analysis of residual solvents in drug substances and products according to ICH, Ph. Eur. and USP
  • An application test is performed for every batch produced
  • Specified concentrations of residual solvents as defined by the ICH
  • SupraSolv® headspace solvents are now available in 500 ml pack sizes, with or without SeccoSept® septum seal cap
  • SeccoSept® protects the quality of these products even better from potential contaminants. It has outstanding self-sealing properties to keep the solvents in flawless condition before, during and after removal
SupraSolv® for headspace gas chromatography Pk Cat. No. Promo price Quantity
Benzyl alcohol 1 l 1.02695.1000E
Benzyl alcohol 2,5 l 1.02695.2500E
Dimethyl sulphoxide (glass bottle) 500 ml 1.01900.0500E
Dimethyl sulphoxide 500 ml 1.01900.0501E
Dimethyl sulphoxide 1 l 1.01900.1000E
Dimethyl sulphoxide 2,5 l 1.01900.2500E
N-Methyl-2-pyrrolidone (NMP) 500 ml 1.02497.0500E
N-Methyl-2-pyrrolidone (NMP) 1 l 1.02497.1000E
N-Methyl-2-pyrrolidone (NMP) 2,5 l 1.02497.2500E
N,N-Dimethylacetamide 500 ml 1.00399.0500E
N,N-Dimethylacetamide 1 l 1.00399.1000E
N,N-Dimethylformamide (glass bottle) 500 ml 1.00202.0500E
N,N-Dimethylformamide 500 ml 1.00202.0501E
N,N-Dimethylformamide 1l 1.00202.1000E
N,N-Dimethylformamide 2,5l 1.00202.2500E
Water for gas chromatography MS 1 l 1.03702.1000E
Water for gas chromatography MS 2,5 l 1.03702.2500E
Water for gas chromatography 1 l 1.02699.1000E
Water for gas chromatography 2,5 l 1.02699.2500E