Would you like to log in again?
Merck special staining kits
Merck special staining kits
VALID UNTIL: 31/10/2017
The reliable solution for critical staining applications
Merck staining kits guarantee high staining reproducibility, since batch-to-batch consistency is verified by chemical, physical and application tests in quality control. Protocols are designed according to efficiency and easy handling.
  • Excellent batch-to-batch consistency with detailed step-by-step protocols
  • Safe packaging prevents cross-contamination during handling
  • Easy handling and read-out after staining
  • High capacity
  • Suitable for IVD applications
Description Average no. of stainings Pk Cat. No. Promo price Quantity
Hematognost FE® staining kit for the detection of free ionic iron (Fe3+) in cells 110 4x250 ml 1.12084.0001E
Weigerts iron hematoxylin kit for nuclear staining in histology 500 2x500 ml 1.15973.0002E
PAS staining kit for detection of aldehyde & mucosubstances 2000 2x500 ml 1.01646.0001E
Reticulin silver plating kit acc. to Gordon & Sweets 110 8x30 ml 1.00251.0001E
Elastica van Gieson staining kit for connective tissue 500 4x500 ml 1.15974.0002E