Would you like to log in again?
Envirocheck® contact slides
Envirocheck® contact slides
VALID UNTIL: 31/10/2017
The flexible solution for monitoring the hygiene of surfaces and liquids
Envirocheck® contact slides are the ideal tool for fast and safe monitoring of the microbiological contamination of surfaces on production lines and equipment or the hygiene status of liquids. These slides provide semi-quantitative results for 5 different groups of microorganisms.
  • Flexible contact slides allow sampling even in difficult to reach areas
  • Protected against contamination: Safe packaging prevents cross-contamination during handling
  • Easy read out after incubation by checking against comparison chart
Description Pk Cat. No. Promo price Quantity
Contact YM(R) (Yeasts & Moulds) Envirocheck® 10 1.02139.0001E
Contact TVC (Total Viable Counts) Envirocheck® 10 1.02149.0001E
Contact DC (Disinfection Control) Envirocheck® 10 1.02147.0001E
Contact E (Enterobacteriaceae) Envirocheck® 10 1.02137.0001E
Contact C (total coliforms/E. coli) Envirocheck® 10 1.02136.0001E