Would you like to log in again?
Chemizorb® for safe and swift absorption
Special prices!
Chemizorb® for safe and swift absorption
VALID UNTIL: 31/12/2017
Remove spilled liquids in your lab with Chemizorb®
Accidents happen in every lab. With Chemizorb®, you can remove aggressive or unpleasant spilled liquids quickly and safely. These absorbents consist of porous mineral or synthetic copolymers that are chemically inert and are capable of taking up 100 to 400% of their own weight in liquid material.
  • Easy, quick, safe and environment-friendly
  • Doesn’t matter which spillage, we have the right solution
  • The 'all rounder' Chemizorb® powder and granules
  • The 'specialist' Chemizorb® alkalis, acids and HF reagents
  • The 'all-in-one' Chemizorb® mercury set
  • NOW AVAILABLE: New composition Chemizorb® H+ - easier to use with higher absorption capacity
This is not the entire Chemizorb® range, discover more in our webshop.
Description Pk Cat. No. Promo price Quantity
Chemizorb® granules absorbent for spilled liquids 5 kg 1.01568.5000E
Chemizorb® H+ absorbent & neutraliser for spilled acids, with indicator 500 g 1.02491.0500E
Chemizorb® H+ absorbent & neutraliser for spilled acids, with indicator 2,5 kg 1.02491.2500E
Chemizorb® Hg reagents refill pack 1 1.12576.0001E