Would you like to log in again?
Memmert ICO CO₂ Incubators
NEW!
Memmert ICO CO₂ Incubators
VALID UNTIL: 30/11/2017
Safety at all times
The new ICO CO₂ incubator is available in four sizes with chamber volumes of
56, 107, 156 and 241 litres.
  • Standard sterilisation program: 60 minutes at 180 °C (without removing the sensors)
  • Heated inner glass door: No condensation in the interior
  • Battery-buffered ControlCOCKPIT (optional): Operates even during a power failure
  • Easy and thorough cleaning: Round corners as the interior is deep-drawn
  • Reliable data logging of temperature, CO2, O2, humidity - open door time conforms to FDA