Would you like to log in again?
Safety shower test unit - exclusive to VWR
Safety shower test unit - exclusive to VWR
VALID UNTIL: 30/06/2015
A mobile test unit designed for easy and convenient testing of the functionality of emergency showers
  • Can be used for both full body and and handheld eye showers
  • 110 l capacity transparent PVC water reservoir to collect and measure flow
  • Unit on brake castors to move around site easily
  • Built in suction pump quickly empties the tank, ready for a repeat check or moving onto the next shower
Body safety showers should be tested monthly to ensure that they are working correctly in case of emergencies. The required water flow rate of 30 l/min is quickly and effectively measured by placing the test unit under the shower and extending the funnel mechanism to under the shower head to collect the water. Up to 110 l of water can be collected over a set period of time and by reading the volume off the scale and the flow rate is easily calculated. The large funnel extends to 2,3 m and a smaller funnel is included for handheld eye showers. The pump switch is IP 45 protected and there are separate mounts for both the power cord and 2 m drainage hose for strorage when not in use. The unit weighs 40 kg and is 60x 50x173 cm (WxDxH).
Description Cat. No. Price Quantity
Safety shower test unit, 230 V, UK-plug 118-0281E