Would you like to log in again?
KIMTECH SCIENCE* Sterling* and STERLING* Nitrile-XTRA Gloves
Get your FREE gift!*
KIMTECH SCIENCE* Sterling* and STERLING* Nitrile-XTRA Gloves
VALID UNTIL: 31/08/2017
Perfect combination of protection, comfort and tactile sensitivity.
For all-round care of science and the environment, KIMTECH SCIENCE* STERLING* Nitrile Gloves perfectly combine protection, comfort and sustainability.
  • Textured fingertips enhance wet and dry grip, with enhanced tactile sensitivity
  • Stronger than latex with better protection against a wide range of chemicals including cytotoxic drugs
  • Virus protection according to ISO 16604 procedure B:2004
  • Anti-static tested, meeting the requirements defined in EN 1149-5
  • Food Contact Approved
*Only one gift per customer and order
Buy 1 pack from the following table and get a FREE KIMTECH phone holder
Size Colour Length Packed Pk Cat. No. Promo price Quantity
KIMTECH SCIENCE* STERLING Gloves
X-Small Grey 240 mm 150/box 150 112-4831P1
Small Grey 240 mm 150/box 150 112-4832P1
Medium Grey 240 mm 150/box 150 112-4833P1
Large Grey 240 mm 150/box 150 112-4834P1
X-Large Grey 240 mm 150/box 150 112-4835P1
KIMTECH SCIENCE*, STERLING* NITRILE-XTRA* Gloves
X-Small Grey 300 mm 100/box 100 112-1356P1
Small Grey 300 mm 100/box 100 112-1357P1
Medium Grey 300 mm 100/box 100 112-1358P1
Large Grey 300 mm 100/box 100 112-1359P1
X-Large Grey 300 mm 90/box 90 112-1368P1
Buy 10 packs from the following table and get FREE KIMTECH wall clock
Size Colour Length Packed Pk Cat. No. Promo price Quantity
KIMTECH SCIENCE* STERLING Gloves
X-Small Grey 240 mm 150/box 150 112-4831P2
Small Grey 240 mm 150/box 150 112-4832P2
Medium Grey 240 mm 150/box 150 112-4833P2
Large Grey 240 mm 150/box 150 112-4834P2
X-Large Grey 240 mm 150/box 150 112-4835P2
KIMTECH SCIENCE*, STERLING* NITRILE-XTRA* Gloves
X-Small Grey 300 mm 100/box 100 112-1356P2
Small Grey 300 mm 100/box 100 112-1357P2
Medium Grey 300 mm 100/box 100 112-1358P2
Large Grey 300 mm 100/box 100 112-1359P2
X-Large Grey 300 mm 90/box 90 112-1368P2